Product Details

SNP ID
rs202013257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41570652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATTTCCTTGATGGCATGTTCTG[C/T]CCCTTCTAACGTGTGGTCCCCACTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR5006 PubMed Links

Gene Details

Gene
MIR5006
Gene Name
microRNA 5006
There are no transcripts associated with this gene.

Gene
VWA8
Gene Name
von Willebrand factor A domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009814.1 5495 Intron NP_001009814.1
NM_015058.1 5495 Missense Mutation ACA,GCA T1809A NP_055873.1
XM_006719791.3 5495 Missense Mutation ACA,GCA T1454A XP_006719854.1
XM_011535007.2 5495 Intron XP_011533309.1
XM_017020469.1 5495 Missense Mutation ACA,GCA T1748A XP_016875958.1
XM_017020470.1 5495 Intron XP_016875959.1
XM_017020471.1 5495 Intron XP_016875960.1
XM_017020472.1 5495 Missense Mutation ACA,GCA T1063A XP_016875961.1
XM_017020473.1 5495 Missense Mutation ACA,GCA T1046A XP_016875962.1
XM_017020474.1 5495 Intron XP_016875963.1

View Full Product Details