Product Details

SNP ID
rs199838787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:21155728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGCTGACATCTCGGATGTTCTG[G/T]CCATGTTTAAGGAACCTTTTACTGG
Phenotype
MIM: 602949
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SAP18 PubMed Links

Gene Details

Gene
SAP18
Gene Name
Sin3A associated protein 18
There are no transcripts associated with this gene.

Gene
SKA3
Gene Name
spindle and kinetochore associated complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166017.1 1328 Intron NP_001159489.1
NM_145061.5 1328 Silent Mutation GGA,GGC G401G NP_659498.4
XM_005266288.3 1328 Silent Mutation GGA,GGC G319G XP_005266345.1
XM_011534994.1 1328 Silent Mutation GGA,GGC G361G XP_011533296.1

View Full Product Details