Product Details

SNP ID
rs200461576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52416061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACATTCCCACAAGCTTAGCAAA[C/T]TCTTCTGATGTTAGGGATCCCTTTT
Phenotype
MIM: 616821 MIM: 610903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
THSD1 PubMed Links

Gene Details

Gene
THSD1
Gene Name
thrombospondin type 1 domain containing 1
There are no transcripts associated with this gene.

Gene
VPS36
Gene Name
vacuolar protein sorting 36 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282168.1 1241 Silent Mutation GAA,GAG E332E NP_001269097.1
NM_001282169.1 1241 Silent Mutation GAA,GAG E283E NP_001269098.1
NM_016075.3 1241 Silent Mutation GAA,GAG E341E NP_057159.2

View Full Product Details