Product Details

SNP ID
rs199656501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:78615922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTGGTCAGATGGAGAGAGGAG[A/G]AAAAAAGAAGTGGGTTTCCATTCAC
Phenotype
MIM: 615906
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF219 PubMed Links

Gene Details

Gene
RNF219
Gene Name
ring finger protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024546.3 1898 Silent Mutation TTC,TTT F613F NP_078822.3
XM_011535225.1 1898 Silent Mutation TTC,TTT F536F XP_011533527.1
XM_017020743.1 1898 Silent Mutation TTC,TTT F536F XP_016876232.1
Gene
RNF219-AS1
Gene Name
RNF219 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details