Product Details

SNP ID
rs200063032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113325960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACAAACAGGCAGATGAACCAGC[A/G]GGACACCAGCAGCGTCCACAGCACA
Phenotype
MIM: 153330
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRTP1 PubMed Links

Gene Details

Gene
GRTP1
Gene Name
growth hormone regulated TBC protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286732.1 929 Missense Mutation CGC,TGC R232C NP_001273661.1
NM_001286733.1 929 Intron NP_001273662.1
NM_024719.3 929 Missense Mutation CGC,TGC R232C NP_078995.2
XM_005268333.3 929 Missense Mutation CGC,TGC R179C XP_005268390.1
XM_011537521.2 929 Intron XP_011535823.1
XM_011537522.2 929 Intron XP_011535824.1
XM_017020755.1 929 Missense Mutation CGC,TGC R154C XP_016876244.1
XM_017020756.1 929 Missense Mutation CGC,TGC R129C XP_016876245.1
Gene
LAMP1
Gene Name
lysosomal associated membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details