Product Details

SNP ID
rs199712386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24189732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGCAGGATGGCGAAGACAAAGA[A/G]GAGGATGCCGTACAAAGTGTACTGC
Phenotype
MIM: 608193
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPO4 PubMed Links

Gene Details

Gene
IPO4
Gene Name
importin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024658.3 1706 Intron NP_078934.3
Gene
REC8
Gene Name
REC8 meiotic recombination protein
There are no transcripts associated with this gene.

Gene
TM9SF1
Gene Name
transmembrane 9 superfamily member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014842.2 1706 Intron NP_001014842.1
NM_001289006.1 1706 Missense Mutation CTC,TTC L415F NP_001275935.1
NM_006405.6 1706 Missense Mutation CTC,TTC L502F NP_006396.2

View Full Product Details