Product Details

SNP ID
rs201945418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73148093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGGACAACCACCTGAGCAATA[C/T]TGTACGTAGCCAGGTACAGTGTCAG
Phenotype
MIM: 104311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSEN1 PubMed Links

Gene Details

Gene
PSEN1
Gene Name
presenilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000021.3 250 Missense Mutation ACT,ATT T25I NP_000012.1
NM_007318.2 250 Missense Mutation ACT,ATT T25I NP_015557.2
XM_005267864.2 250 Missense Mutation ACT,ATT T25I XP_005267921.1
XM_005267866.1 250 Missense Mutation ACT,ATT T25I XP_005267923.1
XM_011536971.2 250 Missense Mutation ACT,ATT T25I XP_011535273.1
XM_011536972.2 250 Missense Mutation ACT,ATT T25I XP_011535274.1
XM_011536973.1 250 Missense Mutation ACT,ATT T25I XP_011535275.1
XM_011536974.1 250 Missense Mutation ACT,ATT T25I XP_011535276.1

View Full Product Details