Product Details
- SNP ID
-
rs201262081
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:59473085 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACAGCTTTAAAATCACCACATTT[C/T]GCTTCCTGAAAAAAGATGAAGGGAG
- Phenotype
-
MIM: 607970
MIM: 614811
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPR135
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1046701] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPR135
- Gene Name
- G protein-coupled receptor 135
There are no transcripts associated with this gene.
- Gene
- L3HYPDH
- Gene Name
- trans-L-3-hydroxyproline dehydratase
View Full Product Details