Product Details

SNP ID
rs201774774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21092703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCAGCAGCTCCTCCTCCTGGCT[A/G]GAGCCGAAACTGCACAGGCCGCACT
Phenotype
MIM: 610018 MIM: 605036
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF40 PubMed Links

Gene Details

Gene
ARHGEF40
Gene Name
Rho guanine nucleotide exchange factor 40
There are no transcripts associated with this gene.

Gene
TMEM253
Gene Name
transmembrane protein 253
There are no transcripts associated with this gene.

Gene
ZNF219
Gene Name
zinc finger protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101672.1 762 Silent Mutation TCC,TCT S198S NP_001095142.1
NM_001102454.1 762 Silent Mutation TCC,TCT S198S NP_001095924.1
NM_016423.2 762 Silent Mutation TCC,TCT S198S NP_057507.2
XM_005267739.1 762 Silent Mutation TCC,TCT S244S XP_005267796.1
XM_006720163.2 762 Silent Mutation TCC,TCT S198S XP_006720226.1
XM_006720164.3 762 Silent Mutation TCC,TCT S198S XP_006720227.1
XM_011536825.1 762 Silent Mutation TCC,TCT S235S XP_011535127.1
XM_017021354.1 762 Silent Mutation TCC,TCT S198S XP_016876843.1
XM_017021355.1 762 Silent Mutation TCC,TCT S198S XP_016876844.1

View Full Product Details