Product Details
- SNP ID
-
rs199907192
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20584109 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGCGCATCACTTCTGTGGAGCT[A/G]CGGATGAAGTTATTGCTCCACTTGC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC254028
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1429539] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC254028
- Gene Name
- uncharacterized LOC254028
There are no transcripts associated with this gene.
- Gene
- RNASE11
- Gene Name
- ribonuclease A family member 11 (inactive)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145250.3 |
550 |
Silent Mutation |
CGC,CGT |
R122R |
NP_660293.1 |
- Gene
- RNASE12
- Gene Name
- ribonuclease A family member 12 (inactive)
There are no transcripts associated with this gene.
View Full Product Details