Product Details

SNP ID
rs201489307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:32092538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACAATCCACTGATGATGAGTA[C/T]GCCTTAGATGGAAAAATTTATGAAC
Phenotype
MIM: 602680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP5 PubMed Links

Gene Details

Gene
ARHGAP5
Gene Name
Rho GTPase activating protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030055.1 2153 Silent Mutation TAC,TAT Y623Y NP_001025226.1
NM_001173.2 2153 Silent Mutation TAC,TAT Y623Y NP_001164.2
XM_005267635.3 2153 Silent Mutation TAC,TAT Y623Y XP_005267692.1
XM_005267636.3 2153 Silent Mutation TAC,TAT Y623Y XP_005267693.1
XM_017021287.1 2153 Silent Mutation TAC,TAT Y623Y XP_016876776.1
XM_017021288.1 2153 Silent Mutation TAC,TAT Y623Y XP_016876777.1

View Full Product Details