Product Details

SNP ID
rs200124758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60245933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGAAAAGAAGAATGGGGAAGA[A/G]AAGGGCAAAGAAGGCAAAAAGAGAA
Phenotype
MIM: 606108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927702 PubMed Links

Gene Details

Gene
LOC101927702
Gene Name
uncharacterized LOC101927702
There are no transcripts associated with this gene.

Gene
PPM1A
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021003.4 177 Intron NP_066283.1
NM_177951.2 177 Intron NP_808820.1
NM_177952.2 177 Missense Mutation AAA,AGA K29R NP_808821.2
XM_005267779.1 177 UTR 5 XP_005267836.1
XM_005267781.1 177 Intron XP_005267838.1
XM_011536872.1 177 Intron XP_011535174.1
XM_011536876.1 177 Intron XP_011535178.1
XM_011536877.1 177 Intron XP_011535179.1
XM_011536878.1 177 Intron XP_011535180.1
XM_011536879.2 177 Intron XP_011535181.1
XM_011536880.2 177 Intron XP_011535182.1
XM_011536881.2 177 Intron XP_011535183.1
XM_011536882.2 177 Intron XP_011535184.1
XM_011536883.1 177 Intron XP_011535185.1
XM_011536884.1 177 Intron XP_011535186.1
XM_017021381.1 177 Intron XP_016876870.1
XM_017021382.1 177 Intron XP_016876871.1
XM_017021383.1 177 Intron XP_016876872.1
XM_017021384.1 177 Intron XP_016876873.1
XM_017021385.1 177 Intron XP_016876874.1
XM_017021386.1 177 Intron XP_016876875.1
XM_017021387.1 177 Intron XP_016876876.1

View Full Product Details