Product Details

SNP ID
rs200233020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77475627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTGGCCTCCTGCAACTGTGCTA[C/G]GTACTCCTCCTCCAGGGGGTTGTCG
Phenotype
MIM: 608466 MIM: 612684
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AHSA1 PubMed Links

Gene Details

Gene
AHSA1
Gene Name
activator of Hsp90 ATPase activity 1
There are no transcripts associated with this gene.

Gene
ISM2
Gene Name
isthmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182509.3 1648 UTR 3 NP_872315.2
NM_199296.2 1648 Missense Mutation CTA,GTA L562V NP_954993.1
XM_011536489.1 1648 Missense Mutation CTA,GTA L474V XP_011534791.1

View Full Product Details