Product Details

SNP ID
rs201601708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35123550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACAGATGAATTCTCAAACTGAAG[A/G]TCATGCCTTGGCACCTGTGAGGAAC
Phenotype
MIM: 609947 MIM: 615902
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0391 PubMed Links

Gene Details

Gene
KIAA0391
Gene Name
KIAA0391
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256678.1 686 Missense Mutation GAT,GGT D102G NP_001243607.1
NM_001256679.1 686 Missense Mutation GAT,GGT D7G NP_001243608.1
NM_001256680.1 686 Intron NP_001243609.1
NM_001256681.1 686 Intron NP_001243610.1
NM_014672.3 686 Missense Mutation GAT,GGT D102G NP_055487.2
XM_005268237.3 686 Missense Mutation GAT,GGT D102G XP_005268294.1
XM_011537409.2 686 Missense Mutation GAT,GGT D102G XP_011535711.1
XM_011537410.2 686 Missense Mutation GAT,GGT D102G XP_011535712.1
XM_017021835.1 686 Missense Mutation GAT,GGT D102G XP_016877324.1
XM_017021836.1 686 Missense Mutation GAT,GGT D102G XP_016877325.1
XM_017021837.1 686 Missense Mutation GAT,GGT D102G XP_016877326.1
Gene
PPP2R3C
Gene Name
protein phosphatase 2 regulatory subunit B''gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305155.1 686 Intron NP_001292084.1
NM_001305156.1 686 Intron NP_001292085.1
NM_017917.3 686 Intron NP_060387.2

View Full Product Details