Product Details

SNP ID
rs200019701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24440415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGAAGAATTGGTGGCCTGAGC[C/G]GATGGGGCCCCCCAGGCCCAGGCGA
Phenotype
MIM: 616162
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KHNYN PubMed Links

Gene Details

Gene
KHNYN
Gene Name
KH and NYN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290256.1 542 UTR 3 NP_001277185.1
NM_001290257.1 542 UTR 3 NP_001277186.1
NM_015299.2 542 UTR 3 NP_056114.1
XM_005267474.4 542 UTR 3 XP_005267531.1
XM_011536590.2 542 UTR 3 XP_011534892.1
XM_017021114.1 542 Intron XP_016876603.1
Gene
LOC101927045
Gene Name
uncharacterized LOC101927045
There are no transcripts associated with this gene.

Gene
SDR39U1
Gene Name
short chain dehydrogenase/reductase family 39U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290292.1 542 Missense Mutation CGC,GGC R102G NP_001277221.1
NM_001290293.2 542 Missense Mutation CGC,GGC R76G NP_001277222.1
NM_001308075.1 542 Missense Mutation CGC,GGC R102G NP_001295004.1
NM_020195.2 542 Missense Mutation CGC,GGC R184G NP_064580.2
XM_011536975.1 542 Missense Mutation CGC,GGC R207G XP_011535277.1
XM_011536976.1 542 Missense Mutation CGC,GGC R142G XP_011535278.1

View Full Product Details