Product Details

SNP ID
rs201908822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60245872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTCTGTTCTGGGAGAAAATGG[C/G]TAGCAGAAGCTACAATTTGTACTAA
Phenotype
MIM: 606108
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927702 PubMed Links

Gene Details

Gene
LOC101927702
Gene Name
uncharacterized LOC101927702
There are no transcripts associated with this gene.

Gene
PPM1A
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021003.4 116 Intron NP_066283.1
NM_177951.2 116 Intron NP_808820.1
NM_177952.2 116 Missense Mutation CTA,GTA L9V NP_808821.2
XM_005267779.1 116 UTR 5 XP_005267836.1
XM_005267781.1 116 Intron XP_005267838.1
XM_011536872.1 116 Intron XP_011535174.1
XM_011536876.1 116 Intron XP_011535178.1
XM_011536877.1 116 Intron XP_011535179.1
XM_011536878.1 116 Intron XP_011535180.1
XM_011536879.2 116 Intron XP_011535181.1
XM_011536880.2 116 Intron XP_011535182.1
XM_011536881.2 116 Intron XP_011535183.1
XM_011536882.2 116 Intron XP_011535184.1
XM_011536883.1 116 Intron XP_011535185.1
XM_011536884.1 116 Intron XP_011535186.1
XM_017021381.1 116 Intron XP_016876870.1
XM_017021382.1 116 Intron XP_016876871.1
XM_017021383.1 116 Intron XP_016876872.1
XM_017021384.1 116 Intron XP_016876873.1
XM_017021385.1 116 Intron XP_016876874.1
XM_017021386.1 116 Intron XP_016876875.1
XM_017021387.1 116 Intron XP_016876876.1

View Full Product Details