Product Details

SNP ID
rs200994696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20999540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTACTCCCATCTGCTTCAAATGG[C/T]TCCAGATTGATGCAGCCAGAGGTAT
Phenotype
MIM: 616091 MIM: 612166
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929718 PubMed Links

Gene Details

Gene
LOC101929718
Gene Name
uncharacterized LOC101929718
There are no transcripts associated with this gene.

Gene
METTL17
Gene Name
methyltransferase like 17
There are no transcripts associated with this gene.

Gene
SLC39A2
Gene Name
solute carrier family 39 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256588.1 286 Missense Mutation CTC,TTC L32F NP_001243517.1
NM_014579.3 286 Missense Mutation CTC,TTC L32F NP_055394.2

View Full Product Details