Product Details

SNP ID
rs201065840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60246012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATGAGAAAAGAGGAAACCAAAT[A/G]AAGAGGATGTGTGAGAGAAAAAAAT
Phenotype
MIM: 606108
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927702 PubMed Links

Gene Details

Gene
LOC101927702
Gene Name
uncharacterized LOC101927702
There are no transcripts associated with this gene.

Gene
PPM1A
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021003.4 256 Intron NP_066283.1
NM_177951.2 256 Intron NP_808820.1
NM_177952.2 256 Missense Mutation ATA,ATG I55M NP_808821.2
XM_005267779.1 256 UTR 5 XP_005267836.1
XM_005267781.1 256 Intron XP_005267838.1
XM_011536872.1 256 Intron XP_011535174.1
XM_011536876.1 256 Intron XP_011535178.1
XM_011536877.1 256 Intron XP_011535179.1
XM_011536878.1 256 Intron XP_011535180.1
XM_011536879.2 256 Intron XP_011535181.1
XM_011536880.2 256 Intron XP_011535182.1
XM_011536881.2 256 Intron XP_011535183.1
XM_011536882.2 256 Intron XP_011535184.1
XM_011536883.1 256 Intron XP_011535185.1
XM_011536884.1 256 Intron XP_011535186.1
XM_017021381.1 256 Intron XP_016876870.1
XM_017021382.1 256 Intron XP_016876871.1
XM_017021383.1 256 Intron XP_016876872.1
XM_017021384.1 256 Intron XP_016876873.1
XM_017021385.1 256 Intron XP_016876874.1
XM_017021386.1 256 Intron XP_016876875.1
XM_017021387.1 256 Intron XP_016876876.1

View Full Product Details