Product Details

SNP ID
rs202087287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105529427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGCTGGTGCTGCCGTGCGTGG[C/T]GCTCAGCGAGGTCAGCATGCAGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105370697 PubMed Links

Gene Details

Gene
LOC105370697
Gene Name
uncharacterized LOC105370697
There are no transcripts associated with this gene.

Gene
TMEM121
Gene Name
transmembrane protein 121
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025268.2 1351 Missense Mutation GCG,GTG A198V NP_079544.1
XM_005268101.3 1351 Missense Mutation GCG,GTG A198V XP_005268158.1
XM_006720261.3 1351 Missense Mutation GCG,GTG A198V XP_006720324.1

View Full Product Details