Product Details

SNP ID
rs202131879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103520153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGCTTCTGAAGTCGCAGCCGCTT[A/C]AGCACCTCCGAGAACTTCTCATGCT
Phenotype
MIM: 123280
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CKB PubMed Links

Gene Details

Gene
CKB
Gene Name
creatine kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001823.4 1088 Silent Mutation CTG,CTT L312L NP_001814.2
XM_017020951.1 1088 Silent Mutation CTG,CTT L336L XP_016876440.1
XM_017020952.1 1088 Intron XP_016876441.1
Gene
TRMT61A
Gene Name
tRNA methyltransferase 61A
There are no transcripts associated with this gene.

View Full Product Details