Product Details

SNP ID
rs199605460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24631947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGTAAGTCAGATTCGCACTTTC[A/G]ATCTTCCTGCACTGTCATCTTCACC
Phenotype
MIM: 123910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GZMB PubMed Links

Gene Details

Gene
GZMB
Gene Name
granzyme B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004131.4 561 Nonsense Mutation CGA,TGA R171* NP_004122.2
XM_011536685.2 561 Nonsense Mutation CGA,TGA R159* XP_011534987.1

View Full Product Details