Product Details
- SNP ID
-
rs201895318
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:65667578 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGATGGTCCACAAAGTCTGCTAC[C/T]GTTATTGCATGATCACCTTCATTTT
- Phenotype
-
MIM: 600382
MIM: 603617
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DENND4A
PubMed Links
Gene Details
- Gene
- DENND4A
- Gene Name
- DENN domain containing 4A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144823.2 |
5358 |
Silent Mutation |
ACA,ACG |
T1703T |
NP_001138295.1 |
NM_001320835.1 |
5358 |
Silent Mutation |
ACA,ACG |
T1704T |
NP_001307764.1 |
NM_005848.4 |
5358 |
Silent Mutation |
ACA,ACG |
T1660T |
NP_005839.3 |
XM_005254120.4 |
5358 |
Silent Mutation |
ACA,ACG |
T1704T |
XP_005254177.1 |
XM_005254121.3 |
5358 |
Silent Mutation |
ACA,ACG |
T1660T |
XP_005254178.1 |
XM_005254122.4 |
5358 |
Silent Mutation |
ACA,ACG |
T970T |
XP_005254179.1 |
XM_006720371.3 |
5358 |
Silent Mutation |
ACA,ACG |
T1704T |
XP_006720434.1 |
XM_011521156.2 |
5358 |
Intron |
|
|
XP_011519458.1 |
XM_011521157.2 |
5358 |
Silent Mutation |
ACA,ACG |
T1188T |
XP_011519459.1 |
XM_017021862.1 |
5358 |
Silent Mutation |
ACA,ACG |
T1704T |
XP_016877351.1 |
XM_017021863.1 |
5358 |
Silent Mutation |
ACA,ACG |
T1660T |
XP_016877352.1 |
XM_017021864.1 |
5358 |
Silent Mutation |
ACA,ACG |
T1704T |
XP_016877353.1 |
XM_017021865.1 |
5358 |
Silent Mutation |
ACA,ACG |
T926T |
XP_016877354.1 |
- Gene
- SLC24A1
- Gene Name
- solute carrier family 24 member 1
There are no transcripts associated with this gene.
View Full Product Details