Product Details
- SNP ID
-
rs201192620
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:29705590 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGCAGACCTTGAAATAGATTTCC[C/T]GCTCAACTCCTTCGGGAATGGCTCC
- Phenotype
-
MIM: 601009
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TJP1
PubMed Links
Gene Details
- Gene
- TJP1
- Gene Name
- tight junction protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301025.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1736R |
NP_001287954.1 |
NM_001301026.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1593R |
NP_001287955.1 |
NM_003257.4 |
5127 |
Missense Mutation |
CAG,CGG |
Q1669R |
NP_003248.3 |
NM_175610.3 |
5127 |
Missense Mutation |
CAG,CGG |
Q1589R |
NP_783297.2 |
XM_005254617.3 |
5127 |
Missense Mutation |
CAG,CGG |
Q1762R |
XP_005254674.2 |
XM_005254618.3 |
5127 |
Missense Mutation |
CAG,CGG |
Q1669R |
XP_005254675.1 |
XM_005254619.3 |
5127 |
Missense Mutation |
CAG,CGG |
Q1682R |
XP_005254676.2 |
XM_005254620.3 |
5127 |
Missense Mutation |
CAG,CGG |
Q1682R |
XP_005254677.2 |
XM_011521972.2 |
5127 |
Missense Mutation |
CAG,CGG |
Q1762R |
XP_011520274.2 |
XM_017022521.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1779R |
XP_016878010.1 |
XM_017022522.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1779R |
XP_016878011.1 |
XM_017022523.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1779R |
XP_016878012.1 |
XM_017022524.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1699R |
XP_016878013.1 |
XM_017022525.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1690R |
XP_016878014.1 |
XM_017022526.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1686R |
XP_016878015.1 |
XM_017022527.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1684R |
XP_016878016.1 |
XM_017022528.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1673R |
XP_016878017.1 |
XM_017022529.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1669R |
XP_016878018.1 |
XM_017022530.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1589R |
XP_016878019.1 |
XM_017022531.1 |
5127 |
Missense Mutation |
CAG,CGG |
Q1353R |
XP_016878020.1 |
View Full Product Details