Product Details

SNP ID
rs201639726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78003335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTCATAAAGGAAAGAGTCCCA[C/T]ATTTTAAAGAGGATGTCACTAACGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC91450 PubMed Links

Gene Details

Gene
LOC91450
Gene Name
uncharacterized LOC91450
There are no transcripts associated with this gene.

Gene
TBC1D2B
Gene Name
TBC1 domain family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015079.5 2544 Missense Mutation ATA,ATG I848M NP_055894.6
NM_144572.1 2544 Missense Mutation ATA,ATG I848M NP_653173.1
XM_011521387.2 2544 Missense Mutation ATA,ATG I811M XP_011519689.1
XM_017022022.1 2544 Missense Mutation ATA,ATG I618M XP_016877511.1

View Full Product Details