Product Details
- SNP ID
-
rs201718359
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:65118799 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATCCTTGGGGAACGAAGTTGCCT[C/T]TGGGATGATCGGATGGCACCAGGTA
- Phenotype
-
MIM: 608138
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PDCD7
PubMed Links
Gene Details
- Gene
- PDCD7
- Gene Name
- programmed cell death 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005707.1 |
1431 |
Missense Mutation |
AAA,AGA |
K459R |
NP_005698.1 |
View Full Product Details