Product Details

SNP ID
rs201424632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76216560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTTAAGGCATGATCCTGATT[A/C]ATTTTTTCTTCAATATCTCAGTCAT
Phenotype
MIM: 608053
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ETFA PubMed Links

Gene Details

Gene
ETFA
Gene Name
electron transfer flavoprotein alpha subunit
There are no transcripts associated with this gene.

View Full Product Details