Product Details

SNP ID
rs201355789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63911947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAGGTGCACCTTCTTCATCAG[C/T]GAGCGGGTGAGGTGGTTGCACAGGG
Phenotype
MIM: 616567
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DAPK2 PubMed Links

Gene Details

Gene
DAPK2
Gene Name
death associated protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014326.3 999 Silent Mutation TCA,TCG S331S NP_055141.2
XM_011521413.2 999 Silent Mutation TCA,TCG S331S XP_011519715.1
XM_011521414.1 999 Silent Mutation TCA,TCG S331S XP_011519716.1
XM_011521415.2 999 Intron XP_011519717.1
XM_011521416.2 999 Intron XP_011519718.1
XM_011521417.1 999 Missense Mutation CAC,CGC H316R XP_011519719.1
XM_011521421.2 999 Silent Mutation TCA,TCG S176S XP_011519723.1
XM_017022043.1 999 Intron XP_016877532.1
XM_017022044.1 999 Silent Mutation TCA,TCG S331S XP_016877533.1
XM_017022045.1 999 Missense Mutation CAC,CGC H316R XP_016877534.1
XM_017022046.1 999 Missense Mutation CAC,CGC H316R XP_016877535.1
XM_017022047.1 999 Intron XP_016877536.1
XM_017022048.1 999 Intron XP_016877537.1
XM_017022049.1 999 Intron XP_016877538.1
XM_017022050.1 999 Intron XP_016877539.1
XM_017022051.1 999 Intron XP_016877540.1
XM_017022052.1 999 Intron XP_016877541.1

View Full Product Details