Product Details

SNP ID
rs201917878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78341217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGACCGTGGACGGACGCAAGT[G/T]CAGGGTGAGGCCCCAGAGCCACTAC
Phenotype
MIM: 180230
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRABP1 PubMed Links

Gene Details

Gene
CRABP1
Gene Name
cellular retinoic acid binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004378.2 350 Missense Mutation TGC,TTC C82F NP_004369.1

View Full Product Details