Product Details

SNP ID
rs202137178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75932701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCCAGGAACCCTCTGGATATT[C/G]ATGCCTCGGGTGTGGTTGGACTGTC
Phenotype
MIM: 609096 MIM: 610894
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXO22 PubMed Links

Gene Details

Gene
FBXO22
Gene Name
F-box protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012170.3 916 Intron NP_036302.1
NM_147188.2 916 Missense Mutation CAT,GAT H271D NP_671717.1
XM_011521447.2 916 Missense Mutation CAT,GAT H167D XP_011519749.1
Gene
FBXO22-AS1
Gene Name
FBXO22 antisense RNA 1
There are no transcripts associated with this gene.

Gene
NRG4
Gene Name
neuregulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138573.3 916 Intron NP_612640.1

View Full Product Details