Product Details

SNP ID
rs201506732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44754912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTTTCTAAAGGTATCCACCAAC[C/G]CCTCCTCCAACATCGATCCAGGCGA
Phenotype
MIM: 616017
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRIM69 PubMed Links

Gene Details

Gene
TRIM69
Gene Name
tripartite motif containing 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301144.1 362 Intron NP_001288073.1
NM_001301145.1 362 Intron NP_001288074.1
NM_001301146.1 362 Intron NP_001288075.1
NM_080745.4 362 Intron NP_542783.2
NM_182985.4 362 Missense Mutation CCC,GCC P7A NP_892030.3

View Full Product Details