Product Details

SNP ID
rs201256320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52552015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGGAATGTGGTCACCAGTGAC[A/C]TCCGTCTTATCCGGAGCTGCAGTAG
Phenotype
MIM: 605487
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARPP19 PubMed Links

Gene Details

Gene
ARPP19
Gene Name
cAMP regulated phosphoprotein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306191.1 556 Missense Mutation GAG,GAT E105D NP_001293120.1
NM_001306195.1 556 Intron NP_001293124.1
NM_001306196.1 556 Missense Mutation GAG,GAT E105D NP_001293125.1
NM_006628.5 556 Missense Mutation GAG,GAT E86D NP_006619.1
XM_017021868.1 556 Intron XP_016877357.1
XM_017021869.1 556 Missense Mutation GAG,GAT E86D XP_016877358.1
XM_017021870.1 556 Intron XP_016877359.1
XM_017021871.1 556 Missense Mutation GAG,GAT E70D XP_016877360.1

View Full Product Details