Product Details

SNP ID
rs202141977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34103055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTGCAGAAACCTGAGCTGGAGA[C/T]GTTTTGGTCAACAAGGCCTCTTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC7 PubMed Links

Gene Details

Gene
EMC7
Gene Name
ER membrane protein complex subunit 7
There are no transcripts associated with this gene.

Gene
PGBD4
Gene Name
piggyBac transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152595.4 983 Missense Mutation ACG,ATG T175M NP_689808.2

View Full Product Details