Product Details
- SNP ID
-
rs202144037
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:60005063 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCCAGAGCTCTCCCGAGAAGATG[C/T]TGCCGCTGAGCGAGATCTACAAGTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXB1
PubMed Links
Gene Details
- Gene
- FOXB1
- Gene Name
- forkhead box B1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_012182.2 |
579 |
Silent Mutation |
CTG,TTG |
L34L |
NP_036314.2 |
View Full Product Details