Product Details
- SNP ID
-
rs200648592
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:44669863 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGCTACCTGGCCCAGGGAACCCT[C/T]GATTCGGACCACTGCATGAGAAGAG
- Phenotype
-
MIM: 614661
MIM: 610844
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PATL2
PubMed Links
Gene Details
- Gene
- PATL2
- Gene Name
- PAT1 homolog 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145112.1 |
1413 |
Missense Mutation |
AAG,GAG |
K264E |
NP_001138584.1 |
XM_011521336.2 |
1413 |
Missense Mutation |
AAG,GAG |
K302E |
XP_011519638.2 |
XM_011521337.2 |
1413 |
Missense Mutation |
AAG,GAG |
K299E |
XP_011519639.2 |
XM_011521338.2 |
1413 |
Missense Mutation |
AAG,GAG |
K264E |
XP_011519640.1 |
XM_011521339.2 |
1413 |
Missense Mutation |
AAG,GAG |
K264E |
XP_011519641.1 |
XM_011521340.2 |
1413 |
Missense Mutation |
AAG,GAG |
K264E |
XP_011519642.1 |
XM_011521341.1 |
1413 |
Missense Mutation |
AAG,GAG |
K264E |
XP_011519643.1 |
XM_011521342.2 |
1413 |
Missense Mutation |
AAG,GAG |
K190E |
XP_011519644.1 |
XM_011521343.2 |
1413 |
Missense Mutation |
AAG,GAG |
K180E |
XP_011519645.1 |
XM_011521344.2 |
1413 |
Missense Mutation |
AAG,GAG |
K180E |
XP_011519646.1 |
XM_011521345.2 |
1413 |
Missense Mutation |
AAG,GAG |
K177E |
XP_011519647.1 |
XM_011521346.2 |
1413 |
Missense Mutation |
AAG,GAG |
K157E |
XP_011519648.2 |
XM_011521347.1 |
1413 |
Missense Mutation |
AAG,GAG |
K75E |
XP_011519649.1 |
XM_011521348.2 |
1413 |
Missense Mutation |
AAG,GAG |
K75E |
XP_011519650.1 |
XM_017022000.1 |
1413 |
Missense Mutation |
AAG,GAG |
K302E |
XP_016877489.1 |
XM_017022001.1 |
1413 |
Missense Mutation |
AAG,GAG |
K177E |
XP_016877490.1 |
- Gene
- SPG11
- Gene Name
- spastic paraplegia 11 (autosomal recessive)
There are no transcripts associated with this gene.
View Full Product Details