Product Details

SNP ID
rs199809549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:29268920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCTTGGCCACAAACTTAAGAAC[C/T]TTCATCTTGCTGGTTTCCAGGTTGG
Phenotype
MIM: 608243
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM189A1 PubMed Links

Gene Details

Gene
FAM189A1
Gene Name
family with sequence similarity 189 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015307.1 897 Intron NP_056122.1
XM_011521407.2 897 Intron XP_011519709.1
Gene
NSMCE3
Gene Name
NSE3 homolog, SMC5-SMC6 complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138704.3 897 Silent Mutation AAA,AAG K262K NP_619649.1

View Full Product Details