Product Details

SNP ID
rs200774497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:62164068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCCTTGCGGCTGCGCCCCACC[A/G]CAGCGCTGCATTGCCAACGCGCAGT
Phenotype
MIM: 610344
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2CD4B PubMed Links

Gene Details

Gene
C2CD4B
Gene Name
C2 calcium dependent domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007595.2 1387 Missense Mutation GCG,GTG A306V NP_001007596.2
XM_017022170.1 1387 Missense Mutation GCG,GTG A306V XP_016877659.1

View Full Product Details