Product Details

SNP ID
rs200086657
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43603352 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGAGAGCTCCATCTCTGCAATCT[C/G]GGTTGCAGACCAGGCTGCTGGGAAT
Phenotype
MIM: 123290 MIM: 606440
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CKMT1B PubMed Links
Additional Information
For this assay, SNP(s) [rs2260160,rs28495368] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CKMT1B
Gene Name
creatine kinase, mitochondrial 1B
There are no transcripts associated with this gene.

Gene
STRC
Gene Name
stereocilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153700.2 2570 Missense Mutation CAG,GAG Q1479E NP_714544.1
XM_011521278.2 2570 Intron XP_011519580.1
XM_017021954.1 2570 Intron XP_016877443.1
XM_017021955.1 2570 Intron XP_016877444.1
XM_017021956.1 2570 Intron XP_016877445.1
XM_017021957.1 2570 Intron XP_016877446.1
XM_017021958.1 2570 Intron XP_016877447.1
XM_017021959.1 2570 Intron XP_016877448.1
XM_017021960.1 2570 Intron XP_016877449.1
XM_017021961.1 2570 Intron XP_016877450.1
XM_017021962.1 2570 Missense Mutation CAG,GAG Q854E XP_016877451.1

View Full Product Details