Product Details

SNP ID
rs200453864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75639648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATCGAAGTCATCGCGCTCCTCA[C/T]TGTACTCTAGCACGTGCCGCTTGAT
Phenotype
MIM: 612980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMP3 PubMed Links

Gene Details

Gene
IMP3
Gene Name
IMP3, U3 small nucleolar ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018285.3 676 Missense Mutation AAT,AGT N174S NP_060755.1
Gene
LOC107984799
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
SNX33
Gene Name
sorting nexin 33
There are no transcripts associated with this gene.

View Full Product Details