Product Details

SNP ID
rs201137112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34102997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGACACTGACAATGACGAGCTC[A/G]AAGTCTTTTTTGCAGTAATGTTACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC7 PubMed Links

Gene Details

Gene
EMC7
Gene Name
ER membrane protein complex subunit 7
There are no transcripts associated with this gene.

Gene
PGBD4
Gene Name
piggyBac transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152595.4 925 Missense Mutation AAA,GAA K156E NP_689808.2

View Full Product Details