Product Details

SNP ID
rs199941779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41987267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTCAGACACGAAGGACCGATGG[A/G]TAAGGATTTCTCGGAAAGAATTGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4E PubMed Links

Gene Details

Gene
PLA2G4E
Gene Name
phospholipase A2 group IVE
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206670.1 786 Missense Mutation ACC,ATC T647I NP_001193599.1
XM_011521237.2 786 Missense Mutation ACC,ATC T613I XP_011519539.1
XM_011521238.1 786 Missense Mutation ACC,ATC T271I XP_011519540.1
XM_011521239.1 786 Missense Mutation ACC,ATC T232I XP_011519541.1
Gene
PLA2G4E-AS1
Gene Name
PLA2G4E antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details