Product Details

SNP ID
rs2229570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70388447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGGCACCAGCACGAAGCTGAC[G/A]TTGGCGGGCAGGTTCTTGGCACTCT
Phenotype
MIM: 607188
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ST3GAL2 PubMed Links

Gene Details

Gene
ST3GAL2
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details