Product Details

SNP ID
rs201177833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30006190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGGGACACTGCTGTCCACTCAG[A/G]CTGAGGACAGAGCCCCGGCCGCAGG
Phenotype
MIM: 604567
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOC2A PubMed Links

Gene Details

Gene
DOC2A
Gene Name
double C2 domain alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282062.1 1391 Missense Mutation GCC,GTC A400V NP_001268991.1
NM_001282063.1 1391 Missense Mutation GCC,GTC A400V NP_001268992.1
NM_001282068.1 1391 Missense Mutation GCC,GTC A400V NP_001268997.1
NM_003586.2 1391 Missense Mutation GCC,GTC A400V NP_003577.2
XM_011545975.1 1391 Missense Mutation GCC,GTC A400V XP_011544277.1
XM_017023776.1 1391 Missense Mutation GCC,GTC A400V XP_016879265.1
XM_017023777.1 1391 Missense Mutation GCC,GTC A400V XP_016879266.1
XM_017023778.1 1391 Missense Mutation GCC,GTC A282V XP_016879267.1
Gene
INO80E
Gene Name
INO80 complex subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304562.1 1391 Intron NP_001291491.1
NM_001304563.1 1391 Intron NP_001291492.1
NM_173618.2 1391 Intron NP_775889.1
XM_011545809.2 1391 Intron XP_011544111.1
XM_011545811.2 1391 Intron XP_011544113.1
XM_011545812.2 1391 Intron XP_011544114.1
XM_017023169.1 1391 Intron XP_016878658.1

View Full Product Details