Product Details

SNP ID
rs201445684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2966224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGCGCTACTGCGACATCCCCTC[C/T]TGTCACAGTGAGTAGCGCGGCTGGA
Phenotype
MIM: 609899 MIM: 614578 MIM: 602474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KREMEN2 PubMed Links

Gene Details

Gene
KREMEN2
Gene Name
kringle containing transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253725.1 659 Silent Mutation TCC,TCT S118S NP_001240654.1
NM_001253726.1 659 Silent Mutation TCC,TCT S118S NP_001240655.1
NM_024507.3 659 Silent Mutation TCC,TCT S118S NP_078783.1
NM_172229.2 659 Silent Mutation TCC,TCT S118S NP_757384.1
Gene
PAQR4
Gene Name
progestin and adipoQ receptor family member 4
There are no transcripts associated with this gene.

Gene
PKMYT1
Gene Name
protein kinase, membrane associated tyrosine/threonine 1
There are no transcripts associated with this gene.

View Full Product Details