Product Details

SNP ID
rs201343471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2165930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCTTTTCAGCCGTCACCACCATC[A/G]CAAAAGGTGAGCCCTTAAGCCAAGG
Phenotype
MIM: 606692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAF7 PubMed Links

Gene Details

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032271.2 274 Missense Mutation ACA,GCA T45A NP_115647.2
XM_005255627.4 274 Missense Mutation ACA,GCA T45A XP_005255684.1
XM_011522700.1 274 Missense Mutation ACA,GCA T16A XP_011521002.1

View Full Product Details