Product Details

SNP ID
rs201023676
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67675387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGGCGCAAGGCCTGCACCATG[C/T]AGACCATGCCCTTCAGGTACAGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf86 PubMed Links

Gene Details

Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
GFOD2
Gene Name
glucose-fructose oxidoreductase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243650.1 964 Intron NP_001230579.1
NM_030819.3 964 Missense Mutation TAC,TGC Y309C NP_110446.3
XM_006721288.3 964 Missense Mutation TAC,TGC Y204C XP_006721351.1
XM_017023738.1 964 Missense Mutation TAC,TGC Y309C XP_016879227.1

View Full Product Details