Product Details

SNP ID
rs200369659
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56653052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAAGGCTCATCAGAGAAGTGCC[A/G]CTGCTGTGCCTGATGTTGGGAGAGC
Phenotype
MIM: 156349 MIM: 156352
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MT1B PubMed Links
Additional Information
For this assay, SNP(s) [rs8047718] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MT1B
Gene Name
metallothionein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005947.2 228 Missense Mutation CAC,CGC H58R NP_005938.1
Gene
MT1DP
Gene Name
metallothionein 1D, pseudogene
There are no transcripts associated with this gene.

Gene
MT1F
Gene Name
metallothionein 1F
There are no transcripts associated with this gene.

View Full Product Details