Product Details
- SNP ID
-
rs199753421
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:15598977 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGGCGCACACAGGAGGTCGACGG[A/G]GGAGTCGTCGGTCAGGCGGAGAAGC
- Phenotype
-
MIM: 614593
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C16orf45
PubMed Links
Gene Details
- Gene
- C16orf45
- Gene Name
- chromosome 16 open reading frame 45
There are no transcripts associated with this gene.
- Gene
- KIAA0430
- Gene Name
- KIAA0430
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184998.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1621S |
NP_001171927.1 |
NM_001184999.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1618S |
NP_001171928.1 |
NM_014647.3 |
4902 |
Missense Mutation |
CCC,TCC |
P1621S |
NP_055462.2 |
XM_005255704.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1622S |
XP_005255761.1 |
XM_005255706.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1620S |
XP_005255763.1 |
XM_005255707.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1612S |
XP_005255764.1 |
XM_017023900.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1622S |
XP_016879389.1 |
XM_017023901.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1621S |
XP_016879390.1 |
XM_017023902.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1568S |
XP_016879391.1 |
XM_017023903.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1479S |
XP_016879392.1 |
XM_017023904.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1478S |
XP_016879393.1 |
XM_017023905.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1443S |
XP_016879394.1 |
XM_017023906.1 |
4902 |
Missense Mutation |
CCC,TCC |
P1442S |
XP_016879395.1 |
View Full Product Details