Product Details

SNP ID
rs200141222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2168089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCCTTGCAGCTGACGGGACCA[A/G]CACCTACAAGCAGCACTGCAGGACA
Phenotype
MIM: 612184 MIM: 606692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASKIN1 PubMed Links

Gene Details

Gene
CASKIN1
Gene Name
CASK interacting protein 1
There are no transcripts associated with this gene.

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032271.2 293 Missense Mutation AAC,AGC N51S NP_115647.2
XM_005255627.4 293 Missense Mutation AAC,AGC N51S XP_005255684.1
XM_011522700.1 293 Missense Mutation AAC,AGC N22S XP_011521002.1

View Full Product Details