Product Details

SNP ID
rs199980902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89686778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATTCGTAAGGAGGGCTTCTTCA[C/T]GGTGCCTCCGGAACACAGGGTGCGC
Phenotype
MIM: 603464
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK10 PubMed Links

Gene Details

Gene
CDK10
Gene Name
cyclin dependent kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098533.2 115 Intron NP_001092003.2
NM_001160367.1 115 Intron NP_001153839.1
NM_052987.3 115 Intron NP_443713.2
NM_052988.4 115 Missense Mutation ACG,ATG T23M NP_443714.3
XM_006721308.2 115 Missense Mutation ACG,ATG T23M XP_006721371.1
XM_006721310.2 115 Missense Mutation ACG,ATG T23M XP_006721373.1
XM_011523405.2 115 Missense Mutation ACG,ATG T23M XP_011521707.1
XM_011523406.2 115 Missense Mutation ACG,ATG T23M XP_011521708.1
XM_011523407.2 115 Missense Mutation ACG,ATG T23M XP_011521709.1
XM_011523408.2 115 Missense Mutation ACG,ATG T23M XP_011521710.1
XM_011523410.2 115 Intron XP_011521712.1
XM_011523411.2 115 UTR 5 XP_011521713.1
XM_011523412.1 115 Intron XP_011521714.1
XM_011523413.2 115 Intron XP_011521715.1
XM_011523414.2 115 Intron XP_011521716.1
XM_011523415.2 115 UTR 5 XP_011521717.1
XM_011523416.2 115 UTR 5 XP_011521718.1
XM_011523417.2 115 Missense Mutation ACG,ATG T23M XP_011521719.1
XM_017023806.1 115 Intron XP_016879295.1
XM_017023807.1 115 Missense Mutation ACG,ATG T23M XP_016879296.1
XM_017023808.1 115 UTR 5 XP_016879297.1
XM_017023809.1 115 UTR 5 XP_016879298.1
XM_017023810.1 115 Intron XP_016879299.1
Gene
SPATA2L
Gene Name
spermatogenesis associated 2 like
There are no transcripts associated with this gene.

View Full Product Details