Product Details

SNP ID
rs200604856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57372595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCCGATATCTCTGTCGTGGCT[G/T]CTCCGCTTGGCCACCTTCTGCCATC
Phenotype
MIM: 602957 MIM: 601880
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCL22 PubMed Links

Gene Details

Gene
CCL22
Gene Name
C-C motif chemokine ligand 22
There are no transcripts associated with this gene.

Gene
CX3CL1
Gene Name
C-X3-C motif chemokine ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304392.1 135 UTR 5 NP_001291321.1
NM_002996.4 135 Silent Mutation CTG,CTT L9L NP_002987.1

View Full Product Details